Masterflex® Spring-Loaded Direct-Read Variable-Area Flowmeter, Air/Water/N2, Dual Scale, Avantor®
Toimittaja: Avantor Fluid Handling
32211-00 32211-02 32211-04 32211-08 32211-16 32211-18 32211-22 32211-24 32211-56 32211-58 32211-60 32211-76 32213-00 32213-10
MFLX32211-12EA 473 EUR
MFLX32211-12 MFLX32211-26 MFLX32213-26 MFLX32211-00 MFLX32211-02 MFLX32211-04 MFLX32211-08 MFLX32211-16 MFLX32211-18 MFLX32211-22 MFLX32211-24 MFLX32211-56 MFLX32211-58 MFLX32211-60 MFLX32211-76 MFLX32213-00 MFLX32213-10
Masterflex® Spring-Loaded Direct-Read Variable-Area Flowmeter, Air/Water/N2, Dual Scale, Avantor®
Virtausmittarit
Perfect low-cost durable process meter.
  • Spring-loaded indicator allows inline mounting in any position
  • Nonreactive wetted parts are ideal for applications requiring a high level of purity
  • Direct reading—has English and metric scales
Spring-loaded, no-float design means these versatile, economical meters do not depend on gravity for accurate readings. Mount in any position in your flow line. Straight pipe runs are not required before or after the flowmeter. Pressure drop is only 4 psi full scale.
Flowmeters feature a knife-edged piston for easy readingMore Product Information