Masterflex® Direct-Read Variable-Area Flowmeters with Valve for Air, 150-mm, Avantor®
Toimittaja: Avantor Fluid Handling
32003-18 32003-20 32003-24 32003-26 32004-16 32004-26 32005-18 32005-20 32005-24 32033-09 32033-15 32034-08 32034-14 32034-15 32035-15
MFLX32003-00EA 225 EUR
MFLX32003-00 MFLX32003-02 MFLX32003-06 MFLX32003-08 MFLX32003-10 MFLX32003-12 MFLX32003-14 MFLX32003-16 MFLX32004-00 MFLX32004-06 MFLX32004-10 MFLX32004-12 MFLX32004-14 MFLX32005-00 MFLX32005-02 MFLX32005-06 MFLX32005-08 MFLX32005-10 MFLX32005-12 MFLX32005-14 MFLX32005-26 MFLX32033-00 MFLX32033-08 MFLX32033-10 MFLX32033-14 MFLX32033-16 MFLX32034-00 MFLX32034-09 MFLX32034-10 MFLX32034-16 MFLX32035-00 MFLX32035-08 MFLX32035-09 MFLX32035-10 MFLX32035-14 MFLX32035-16 MFLX32003-18 MFLX32003-20 MFLX32003-24 MFLX32003-26 MFLX32004-16 MFLX32004-26 MFLX32005-18 MFLX32005-20 MFLX32005-24 MFLX32033-09 MFLX32033-15 MFLX32034-08 MFLX32034-14 MFLX32034-15 MFLX32035-15
Masterflex® Direct-Read Variable-Area Flowmeters with Valve for Air, 150-mm, Avantor®
Virtausmittarit
No more eye strain - front shield magnifies scale 16 percent for easier readings.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility