Masterflex® Direct-Read Variable-Area Flowmeters for Gases, 65-mm, Aluminum, Avantor®
Toimittaja: Avantor Fluid Handling
32006-18 32006-24 32006-26 32006-28 32006-34 32007-14 32007-24 32007-34
MFLX32006-08EA 386 EUR
MFLX32006-08 MFLX32006-10 MFLX32006-12 MFLX32006-14 MFLX32006-16 MFLX32006-30 MFLX32006-32 MFLX32006-36 MFLX32006-38 MFLX32007-30 MFLX32007-32 MFLX32007-38 MFLX32006-18 MFLX32006-24 MFLX32006-26 MFLX32006-28 MFLX32006-34 MFLX32007-14 MFLX32007-24 MFLX32007-34
Masterflex® Direct-Read Variable-Area Flowmeters for Gases, 65-mm, Aluminum, Avantor®
Virtausmittarit
Ideal for gas applications where metal components must be minimized.
  • Transparent front safety shield enhances reading resolution by 16%
  • Rugged design features case-enclosed flowtube
  • Tubes include a fused ceramic scale as a precise, permanent measuring guide
These meters eliminate the need for correlation charts with direct-reading scales for individual routine gases. The inert PTFE and glass wetted parts combine with the rigid anodized aluminum frame to offer a unique solution for ultra-pure gas applications. Flowtubes are replaceable.