Masterflex® Panel-Mount Direct-Read Variable-Area Flowmeters for Air, 150-mm Scale, Avantor®
Toimittaja: Avantor Fluid Handling
32000-00 32000-02 32000-08 32000-18 32000-22 32001-06 32001-08 32001-10 32001-14 32001-16 32001-18 32001-24 32002-08 32002-10 32002-12 32030-00 32030-10 32031-15 32032-00 32032-08 32032-09 32032-10 32032-14 32032-15 32032-16
MFLX32000-06EA 205 EUR
MFLX32000-06 MFLX32000-10 MFLX32000-12 MFLX32000-14 MFLX32001-00 MFLX32001-12 MFLX32002-00 MFLX32002-06 MFLX32002-14 MFLX32030-08 MFLX32030-09 MFLX32030-14 MFLX32030-15 MFLX32030-16 MFLX32031-00 MFLX32031-10 MFLX32000-00 MFLX32000-02 MFLX32000-08 MFLX32000-18 MFLX32000-22 MFLX32001-06 MFLX32001-08 MFLX32001-10 MFLX32001-14 MFLX32001-16 MFLX32001-18 MFLX32001-24 MFLX32002-08 MFLX32002-10 MFLX32002-12 MFLX32030-00 MFLX32030-10 MFLX32031-15 MFLX32032-00 MFLX32032-08 MFLX32032-09 MFLX32032-10 MFLX32032-14 MFLX32032-15 MFLX32032-16
Masterflex® Panel-Mount Direct-Read Variable-Area Flowmeters for Air, 150-mm Scale, Avantor®
Virtausmittarit
No more eye strain—front shield magnifies scale 16 percent for easier readings.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility