Acetal Fitting Manifolds, Female NPT
Toimittaja: Avantor Fluid Handling
31524-16
MFLX31524-16EA 175 EUR
MFLX31524-16
Acetal Fitting Manifolds, Female NPT
Manifoldit
Come in a variety of configurations—choose the number of ports and connections to suit your needs!
  • Expand your flow and line capacity
Constructed of acetal plastic for durable applications and for use with chemically compatible fluids.