PTFE Fitting Manifolds, Female NPT
Toimittaja: Avantor Fluid Handling
31521-01 31521-07 31521-09 31521-11 31521-13 31521-15 31521-17 31521-19 31521-21 31521-25 31521-27
MFLX31521-01EA 130 EUR
MFLX31521-01 MFLX31521-07 MFLX31521-09 MFLX31521-11 MFLX31521-13 MFLX31521-15 MFLX31521-17 MFLX31521-19 MFLX31521-21 MFLX31521-25 MFLX31521-27
PTFE Fitting Manifolds, Female NPT
Manifoldit
Chemically-inert PTFE handles some of your most aggressive chemical solutions.
  • Constructed of durable PTFE
Come in a variety of configurations—choose the number of ports and connections to suit your needs!