Masterflex® Certified Sanitary Adapters, ADCF Polypropylene, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX31515-89EA 78.2 EUR
MFLX31515-89 MFLX31515-90 MFLX31515-93 MFLX31515-95 MFLX31515-97 MFLX31515-99 MFLX31515-91 MFLX31515-96 MFLX31515-92
Masterflex® Certified Sanitary Adapters, ADCF Polypropylene, Avantor®
Kiinnitykset ja liittimet
Certificate of compliance included!
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Animal derivative component free
  • Meet USP Class IV requirements
  • Sterilisable vis gamma irradiation
These sanitary fittings are the perfect to transition from sanitary clamp connections to seal or barb for flexible tubing. All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow - and no particulate build-up. There are no sharp edges in the flow path, so cleaning is very easy. Made from animal-derivedMore Product Information