Adapter Fittings, Compression to NPT(M), Brass
Toimittaja: Avantor Fluid Handling
31412-35 31412-36 31412-69 31412-71 31412-72 31412-75 31412-78 31412-80
MFLX31412-35EA 9.4 EUR
MFLX31412-35 MFLX31412-36 MFLX31412-69 MFLX31412-71 MFLX31412-72 MFLX31412-75 MFLX31412-78 MFLX31412-80
Adapter Fittings, Compression to NPT(M), Brass
Kiinnitykset ja liittimet
Create a secure connection with compression fittings.
  • Constructed of durable brass
Good chemical resistance. Rated to 400 psi maximum at 77 °F ( 25 °C). Usable from -85 to 482 °F (-65 to 250 °C).