CPC® High-Flow Quick-Disconnect Fittings, Hose Barb Inserts
Toimittaja: Avantor Fluid Handling
31303-43 31303-46 31303-43BLK 31303-46BLK
MFLX31303-40EA 11.2 EUR
MFLX31303-40 MFLX31303-41 MFLX31303-42 MFLX31303-45 MFLX31303-47 MFLX31303-48 MFLX31307-40 MFLX31307-41 MFLX31307-45 MFLX31307-46 MFLX31307-48 MFLX31303-43 MFLX31303-46 MFLX31303-43BLK MFLX31303-46BLK MFLX31303-42BLK MFLX31303-47BLK MFLX31303-48BLK MFLX31307-40BLK MFLX31307-41BLK MFLX31307-48BLK MFLX31303-40BLK MFLX31303-41BLK MFLX31303-45BLK MFLX31307-45BLK MFLX31307-46BLK
CPC® High-Flow Quick-Disconnect Fittings, Hose Barb Inserts
Kiinnitykset ja liittimet
No more leaks - valved couplings automatically shut off flow.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Compact, lightweight couplings feature a high flow and low spillage design
These CPC® high-flow quick-disconnect fittings have flow comparable to many ¹/₂" flow couplings in a ³/₈" body size. An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately.More Product Information