CPC® High-Flow Quick-Disconnect Fittings, Threaded Bodies
Toimittaja: Avantor Fluid Handling
31308-29BLK
MFLX31303-00EA 25.3 EUR
MFLX31303-00 MFLX31303-01 MFLX31303-02 MFLX31303-06 MFLX31307-00 MFLX31307-01 MFLX31307-02 MFLX31303-00BLK MFLX31303-06BLK MFLX31303-01BLK MFLX31307-00BLK MFLX31307-01BLK MFLX31307-02BLK MFLX31303-02BLK MFLX31308-29BLK
CPC® High-Flow Quick-Disconnect Fittings, Threaded Bodies
Kiinnitykset ja liittimet
No more leaks - valved couplings automatically shut off flow.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Compact, lightweight couplings feature a high flow and low spillage design
These CPC® high-flow quick-disconnect fittings have flow comparable to many ¹/₂" flow couplings in a ³/₈" body size. An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately.More Product Information