CPC® High-Flow Quick-Disconnect Fittings, Bulkhead Panel Mount Hose Barb Bodies
Toimittaja: Avantor Fluid Handling
31308-36BLK 31308-38BLK 31308-65BLK 31308-67BLK
MFLX31303-20EA 40.3 EUR
MFLX31303-20 MFLX31303-21 MFLX31303-26 MFLX31303-28 MFLX31307-20 MFLX31303-21BLK MFLX31307-20BLK MFLX31303-20BLK MFLX31303-28BLK MFLX31303-26BLK MFLX31308-36BLK MFLX31308-38BLK MFLX31308-65BLK MFLX31308-67BLK
CPC® High-Flow Quick-Disconnect Fittings, Bulkhead Panel Mount Hose Barb Bodies
Kiinnitykset ja liittimet
No more leaks - valved couplings automatically shut off flow.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Compact, lightweight couplings feature a high flow and low spillage design
These CPC® high-flow quick-disconnect fittings have flow comparable to many ¹/₂" flow couplings in a ³/₈" body size. An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately.More Product Information