CPC® High-Flow Quick-Disconnect Fittings, Hose Barb Bodies
Toimittaja: Avantor Fluid Handling
31303-16 31307-13 31307-13BLK 31303-16BLK 31308-34BLK 31308-63BLK
MFLX31303-10EA 25.3 EUR
MFLX31303-10 MFLX31303-11 MFLX31303-12 MFLX31303-13 MFLX31303-15 MFLX31307-10 MFLX31307-11 MFLX31307-15 MFLX31303-16 MFLX31307-13 MFLX31307-13BLK MFLX31303-16BLK MFLX31303-11BLK MFLX31303-10BLK MFLX31303-12BLK MFLX31303-13BLK MFLX31303-15BLK MFLX31307-11BLK MFLX31307-10BLK MFLX31307-15BLK MFLX31308-34BLK MFLX31308-63BLK
CPC® High-Flow Quick-Disconnect Fittings, Hose Barb Bodies
Kiinnitykset ja liittimet
No more leaks - valved couplings automatically shut off flow.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Compact, lightweight couplings feature a high flow and low spillage design
These CPC® high-flow quick-disconnect fittings have flow comparable to many ¹/₂" flow couplings in a ³/₈" body size. An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately.More Product Information