Masterflex® Color-Coded Sanitary Clamp Fittings, Nylon, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX31201-43EA 23.9 EUR
MFLX31201-43 MFLX31201-44 MFLX31201-45 MFLX31201-46 MFLX31201-48
Masterflex® Color-Coded Sanitary Clamp Fittings, Nylon, Avantor®
Kiinnitykset ja liittimet
Color-coded knobs for easy identification.
  • Choose from 4 color options for high visibility
  • Lightweight design puts less stress on your system
  • Lot and batch traceability is ideal for increased security and compliance safety
  • Compatible with ASME-BPE and BPSA standard nonmetallic and stainless steel flange ferrules
Constructed of glass-reinforced nylon, these disposable Tri-Clamp® sanitary clamps are perfect for single-use transfer lines in biopharma markets, plus a wide range of fluid transfer systems. The nylon material meets USP Class VI and FDAMore Product Information