Masterflex® Reducer Fittings, Hose Barb, Straight (Nylon), Avantor®
Toimittaja: Avantor Fluid Handling
MFLX30616-10EA 28 EUR
MFLX30616-10 MFLX30616-30 MFLX30621-97 MFLX30622-02 MFLX30622-04 MFLX30622-10 MFLX30622-14 MFLX30622-18 MFLX30622-25 MFLX30622-28 MFLX30622-30 MFLX30622-34 MFLX30622-39 MFLX30626-10 MFLX30626-28 MFLX30626-34 MFLX30626-43 MFLX30703-99 MFLX30704-89 MFLX31518-39 MFLX40616-02 MFLX40616-06 MFLX40621-94 MFLX40622-20 MFLX40622-22 MFLX41517-92 MFLX41518-28 MFLX30621-99 MFLX30622-06 MFLX30622-08 MFLX30622-12 MFLX30622-16 MFLX30622-41 MFLX30622-43 MFLX30626-04 MFLX30626-06 MFLX30626-08 MFLX30626-16 MFLX30626-30 MFLX30626-39 MFLX30626-41 MFLX31518-27 MFLX31518-32 MFLX31518-35 MFLX31518-37 MFLX40625-94 MFLX40626-20 MFLX40626-22 MFLX41518-30
Masterflex® Reducer Fittings, Hose Barb, Straight (Nylon), Avantor®
Kiinnitykset ja liittimet
Best for low-pressure applications.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Temperture