Gauge Guard Protector and Isolator Seals
Toimittaja: Avantor Fluid Handling
30565-01 30565-21
MFLX30565-19EA 90.5 EUR
MFLX30565-19 MFLX30565-01 MFLX30565-21
Gauge Guard Protector and Isolator Seals
Sulkijakalvot ja -matot Tiivisteet
Extend the life and accuracy of your expensive and fragile instruments.
  • Manufactured to meet USP Class VI certification
  • Animal derived ingredient free
Gauge Guard Protectors safeguard your expensive instruments from damaging process fluids and destructive routine maintenance such as autoclaving. Gauge guard is platinum-cured silicone combined with a 316 stainless steel perforated plate, and slips over the OD of your diaphragm seal. Model 30565-03 fits both 1 and 1 1/2" seals.
Gauge Guard Isolator Seals combine a hygenic seal with a membrane to isolateMore Product Information