Masterflex® Pressure Gauge Guard Gaskets and Protector Caps, Avantor®
Toimittaja: Avantor Fluid Handling
30565-14 30565-88
MFLX30565-02EA 75.9 EUR
MFLX30565-02 MFLX30565-06 MFLX30565-08 MFLX30565-62 MFLX30565-64 MFLX30565-86 MFLX30565-14 MFLX30565-88
Masterflex® Pressure Gauge Guard Gaskets and Protector Caps, Avantor®
Sulkijakalvot ja -matot Tiivisteet
Simple accessories to protect your delicate pressure gauge.
  • Choose from platinum-cured silicone, FKM, EPDM or PTFE materials
Use these gaskets and caps to protect the sensitive pressure membrane on your sanitary pressure gauge! All are available for sanitary clamp connections in sizes from 3/4" to 2".
Gaskets feature sanitary seal edges and a full membrane made of the same material as the sealing gasket edge. No more worry about particulates and film building up on your pressure membrane! Available in platinum-cured silicone, FKM, EPDM and PTFE.