Masterflex® Sanitary Tri-Clamp® Sock Screen Gaskets, Avantor®
Toimittaja: Avantor Fluid Handling
30563-08 30563-18 30563-22 30563-24 30563-26 30563-36 30563-38 30563-40 30563-44 30563-46 30563-48 30563-62 30563-64 30563-76 30563-78
MFLX30563-00EA 178 EUR
MFLX30563-00 MFLX30563-02 MFLX30563-04 MFLX30563-06 MFLX30563-16 MFLX30563-20 MFLX30563-42 MFLX30563-60 MFLX30563-68 MFLX30563-08 MFLX30563-18 MFLX30563-22 MFLX30563-24 MFLX30563-26 MFLX30563-36 MFLX30563-38 MFLX30563-40 MFLX30563-44 MFLX30563-46 MFLX30563-48 MFLX30563-62 MFLX30563-64 MFLX30563-76 MFLX30563-78
Masterflex® Sanitary Tri-Clamp® Sock Screen Gaskets, Avantor®
Sulkijakalvot ja -matot Tiivisteet
Greater particulate capacity for higher particulate flow.
  • Ideal for tank-to-tank transfer lines, inline fluid conditioning, and pump protection
  • Greater capacity than standard screen gaskets, so less down time for the fluid line
Sock screen gaskets extend the run time in your lines between changing out and cleaning standard screen gaskets. Featuring an extended sock-shaped mesh with 300% more surface area, these gaskets are ideal for lines with higher particular counts and greater flow volumes. Gaskets are available in platinum-cured silicone, EPDM,More Product Information