Sanitary Ball Valves with Tri-Clamp Connections
Toimittaja: Avantor Fluid Handling
30530-00 30530-01 30530-02 30530-03
MFLX30530-00EA 195 EUR
MFLX30530-00 MFLX30530-01 MFLX30530-02 MFLX30530-03
Sanitary Ball Valves with Tri-Clamp Connections
Venttiilit Palloventtiilit
Don’t worry about leaks.
  • Wetted parts: 316 stainless steel with a sanitary finish and PTFE seals.
Ball valves feature a unique design that allows for easy in-line servicing. Valve seats are made of fully encapsulated PTFE. Each valve includes a locking handle.