Precision Digital ProtEX Explosion-Proof Process Meters
Toimittaja: Avantor Fluid Handling
30004-68
MFLX30004-68EA 925 EUR
MFLX30004-68
Precision Digital ProtEX Explosion-Proof Process Meters
Valvontajärjestelmät Prosessin seuranta
Four sensors operate as through-glass buttons for easy on-site programming without removing the cover and exposing the electronics.
  • High-contrast backlit LCD is easy to read from a distance and under most lighting conditions
  • 5-digit rate and 7-character total/tag display
  • 32-point, square root, or exponential linearization
  • 90° rotatable display
  • 5-digit password prevents unauthorized changes to the programmed parameter settings
  • Explosion-proof, IP68, NEMA 4X enclosure
  • FM, CSA, ATEX, IECEx, CE approved
The ProtEX PD6800 is a rugged, explosion-proofMore Product Information