LEVIFLOW® Single-Use Clamp-On Ultrasonic Flow Sensors
Toimittaja: Avantor Fluid Handling
MFLX24500-19EA 3160 EUR
MFLX24500-19 MFLX24500-18
LEVIFLOW® Single-Use Clamp-On Ultrasonic Flow Sensors
Fluid Sensors
No moving parts—no particle generation—no contamination.
  • Starter kit and flow sensor required to build a complete system
  • Use on your existing silicone or C-Flex tubing
  • Obtain flow measurement without intrusion
  • Reusable flow sensors—choose from 4-, 20-, or 80-LPM models
These flow systems are ideal for sterile applications. Flow sensors fit over your existing silicone or C-Flex tubing lines and are easily reusable—remove sensor clamp, dispose of flexible tubing, and fix sensor over a new line.
These non-invasive flow measurement systems have no movingMore Product Information