Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Peroxide-Cured Silicone, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX07616-22EA 105 EUR
MFLX07616-22 MFLX07616-24 MFLX07616-26 MFLX07616-28 MFLX07616-30 MFLX07616-32 MFLX07616-34 MFLX07616-36 MFLX07616-38 MFLX07616-40 MFLX07616-42 MFLX07616-44 MFLX07616-46 MFLX07616-48 MFLX07616-12
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Peroxide-Cured Silicone, Avantor®
Letku Pumppuletku
Long service life - outstanding fatigue resistance due to flexing!
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Appearance: Opaque
Temperature: -70 to 52 °C
Life at 100 rpm: 1000 hrs
The first of a series of two-stop extension tubing options is below under Accessories. See all of our two-stopMore Product Information