FMI Series R Valveless Piston Pump Heads
Toimittaja: Avantor Fluid Handling
07117-66 07117-67
MFLX07115-99EA 358 EUR
MFLX07115-99 MFLX07117-99 MFLX07117-66 MFLX07117-67
FMI Series R Valveless Piston Pump Heads
Pumput Pumppupäät Mäntäpumppupäät
Compatible with Masterflex® L/S® or series Q drive.
  • No valves to clog or wear out
  • Use for low-flow applications
  • Kynar® pump body withstands harsh chemicals
  • Series R pump heads come equipped with rotating pistons to prevent clogging—ideal for pumping visco
Compression fittings accept 1/4" OD tubing. For a complete assembly, order series R pump head and a Masterflex L/S® or series Q drive. Adapter kits let you attach pump heads to drives.
Flow characteristics may vary with different fluids.