Flux Pumps Sanitary Drum Pump Air Motors
Toimittaja: Avantor Fluid Handling
07070-60 07070-70
MFLX07070-60EA 1140 EUR
MFLX07070-60 MFLX07070-70
Flux Pumps Sanitary Drum Pump Air Motors
Pumput Rumpupumput
Configurable to your requirements.
  • High-viscosity tube set handles fluids with viscosities to 50,000 cp!
Air-operated motors are ideal for hazardous duty applications or where electricity is not available. Models 07070-60 and -70 are fitted with a muffler and include a grounding post. Model 07070-70 also has a unique trigger valve handle—press switch for intermittent pumping; lock switch for continuous use.