Miniature Gear Pumps - Compact Plastic Head and Drive
Toimittaja: Avantor Fluid Handling
07012-00 07012-02 07012-20 07012-30
MFLX07012-00EA 0 EUR
MFLX07012-00 MFLX07012-02 MFLX07012-20 MFLX07012-30
Miniature Gear Pumps - Compact Plastic Head and Drive
Pumput Hammaspyöräpumput
Complete mini gear pump systems for low-flow applications.
  • Maximum flow rates up to 0.67 GPM
  • 115 VAC, 230 VAC, and 12 VDC models
These miniature gear pumps feature a compact plastic head and drive. Choose from three model options— nonenclosed OEM, lab, and fully enclosed lab versions.
Toimitustiedot: Includes models 07012-00, -02 have 12" (30.4-cm) leads with no plug; models 07012-30, -32 have an aluminum enclosure and a 7-ft (2.1-m) cord with grounded plug and in-line switch; model 07012-20 has an 8-inch (20.3-m) cord with no plug.