Masterflex® Ismatec® Pump Tubing, Versilon™ 2001, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX06518-04EA 118 EUR
MFLX06518-04 MFLX96467-14 MFLX96467-28 MFLX96467-36 MFLX96467-42 MFLX96467-48 MFLX96467-22
Masterflex® Ismatec® Pump Tubing, Versilon™ 2001, Avantor®
Letku Pumppuletku
Compatible with aggressive chemicals – even MEK!
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Outstanding chemical resistance — withstands acetone, DMSO, ketones, and more
  • Cost-effective alternative to fluoropolymers or fluoroelastomers
The Tygon Chemical 2001 translucent tubing hasMore Product Information