Masterflex® L/S® Precision Pump Tubing, Tygon® E-Lab, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX06509-13EA 72.8 EUR
MFLX06509-13 MFLX06509-14 MFLX06509-16 MFLX06509-17 MFLX06509-18 MFLX06509-25
Masterflex® L/S® Precision Pump Tubing, Tygon® E-Lab, Avantor®
Letku Pumppuletku
Ideal for general transfer applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-