Masterflex® Ismatec® Pump Tubing, Tygon® LMT-55, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX06507-01EA 61 EUR
MFLX06507-01 MFLX06507-09 MFLX06507-11 MFLX06507-21 MFLX06507-39 MFLX06507-05 MFLX06507-15
Masterflex® Ismatec® Pump Tubing, Tygon® LMT-55, Avantor®
Letku Pumppuletku
Exceptional durability with long life.
  • Crystal clear and flexible while also being long-lasting and crack-resistant
Applications: General laboratory, food and beverage, biopharmaceutical, analytical instruments
Sterilize by autoclave or ethylene oxide gas.