Masterflex® L/S® Precision Pump Tubing, PharMed® BPT, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX06452-14EA 105 EUR
MFLX06452-14 MFLX06452-16 MFLX06452-13 MFLX06452-17
Masterflex® L/S® Precision Pump Tubing, PharMed® BPT, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Avantor®
Letku Pumppuletku
Protect your light-sensitive samples.
  • Genuine Masterflex® pump tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Gentle, low pulsation and accurate dosing and flow when used in the Masterflex Ultrapharm™ L/S® pump head
  • Up to 10,000 hours of tubing life—reduces tubing costs and pump downtime
  • Resists ozone and UV