Masterflex® B/T® PerfectPosition® Pump Tubing, Tygon® S3™ E-LFL, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX06440-87EA 346 EUR
MFLX06440-87 MFLX06440-91 MFLX06449-87 MFLX06449-91
Masterflex® B/T® PerfectPosition® Pump Tubing, Tygon® S3™ E-LFL, Avantor®
Letku Pumppuletku
Ensure optimal performance from your Masterflex® B/T series pump.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and