Masterflex® L/S® Precision Pump Tubing, C-Flex® ULTRA, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX06434-14EA 85.5 EUR
MFLX06434-14 MFLX06434-17 MFLX06434-25 MFLX06434-16 MFLX06434-18 MFLX06437-16
Masterflex® L/S® Precision Pump Tubing, C-Flex® ULTRA, Avantor®
Letku Pumppuletku
Longest life of any C-Flex® formulation.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-