Masterflex® L/S® High-Performance Precision Pump Tubing, C-Flex®, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX06424-15EA 106 EUR
MFLX06424-15 MFLX06424-24 MFLX06424-35 MFLX06424-36
Masterflex® L/S® High-Performance Precision Pump Tubing, C-Flex®, Avantor®
Letku Pumppuletku
Combines biocompatibility of silicone with chemical resistance similar to Tygon®.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include