Masterflex® I/P® Precision Pump Tubing, Tygon® E-Food, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX06418-26EA 376 EUR
MFLX06418-26 MFLX06418-73 MFLX06418-82
Masterflex® I/P® Precision Pump Tubing, Tygon® E-Food, Avantor®
Letku Pumppuletku
The preferred tubing of the food industry.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory Approvals to match your unique application
  • Smooth, nonporous inner bore inhibits particle entrapment and minimises