Masterflex® Microbore Transfer Tubing, PTFE, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX06417-11EA 34.2 EUR
MFLX06417-11 MFLX06417-21 MFLX06417-31 MFLX06417-41 MFLX06417-51
Masterflex® Microbore Transfer Tubing, PTFE, Avantor®
Letku Siirtoputki
Ideal of extreme temperatures and harsh chemical environments.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Use this translucent tubing in extreme temperatures and in harsh chemical environments. It's chemically inert, has low permeability, and has the lowest friction coefficient of any tubing. TemperatureMore Product Information