Masterflex® I/P® High-Performance Precision Pump Tubing, Versilon™ A-60-G, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX06404-70EA 327 EUR
MFLX06404-70 MFLX06404-88 MFLX06404-89
Masterflex® I/P® High-Performance Precision Pump Tubing, Versilon™ A-60-G, Avantor®
Letku Pumppuletku
Outlast and outperform other tubing with 10,000 hours of tubing life.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory Approvals to match your unique application
  • Opaque beige color protects light and UV sensitive