Masterflex® I/P® Precision Pump Tubing, Tygon® A-60-F, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX06402-26EA 309 EUR
MFLX06402-26 MFLX06402-73 MFLX06402-82
Masterflex® I/P® Precision Pump Tubing, Tygon® A-60-F, Avantor®
Letku Pumppuletku
Stay compliant with food grade applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory Approvals to match your unique application
  • Opaque beige color protects light and UV sensitive samples
Masterflex®More Product Information