Masterflex® L/S® Precision Pump Tubing, Tygon® A-60-F and A-70-F, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX06402-13EA 101 EUR
MFLX06402-13 MFLX06402-14 MFLX06402-16 MFLX06402-17 MFLX06402-18 MFLX06402-25
Masterflex® L/S® Precision Pump Tubing, Tygon® A-60-F and A-70-F, Avantor®
Letku Pumppuletku
Similar to Norprene® but with FDA approval.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-