Masterflex® L/S® High-Performance Precision Pump Tubing, Tygon® Fuel and Lubricant, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX06401-15EA 213 EUR
MFLX06401-15 MFLX06401-24 MFLX06401-35 MFLX06401-36
Masterflex® L/S® High-Performance Precision Pump Tubing, Tygon® Fuel and Lubricant, Avantor®
Letku Pumppuletku
Ideal for transferring hydrocarbons, gasoline, kerosene, heating oils, cutting compounds, and glycol-based coolants.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application