Masterflex® Transfer Tubing, Metric Polytetrafluoroethylene (PTFE), Avantor®
Toimittaja: Avantor Fluid Handling
MFLX06363-30EA 268 EUR
MFLX06363-30 MFLX06363-10 MFLX06363-20 MFLX06605-52 MFLX06605-53 MFLX06605-54 MFLX06605-55 MFLX06605-56 MFLX06605-57
Masterflex® Transfer Tubing, Metric Polytetrafluoroethylene (PTFE), Avantor®
Letku Siirtoputki
Withstands extreme temperatures.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Good for high-purity work. PTFE tubing withstands extreme temperatures.
Characteristics: Chemically inert, nontoxic, andMore Product Information