CPC® Quick-Disconnect Fittings, Threaded Inserts
Toimittaja: Avantor Fluid Handling
06361-81 06361-81BLK
MFLX06361-29EA 13.5 EUR
MFLX06361-29 MFLX06361-31 MFLX06361-65 MFLX06361-91 MFLX06361-81 MFLX06361-81BLK MFLX06361-91BLK MFLX06361-29BLK MFLX06361-31BLK MFLX06361-65BLK
CPC® Quick-Disconnect Fittings, Threaded Inserts
Kiinnitykset ja liittimet
No more struggling - just press thumb latch.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-More Product Information