CPC® Quick-Disconnect Fittings, Threaded Bodies
Toimittaja: Avantor Fluid Handling
MFLX06359-70EA 15 EUR
MFLX06359-70 MFLX06359-71 MFLX06359-72 MFLX06359-73 MFLX06359-80 MFLX06359-81 MFLX06359-82 MFLX06359-83 MFLX06361-00 MFLX06361-42 MFLX06361-43 MFLX06359-71BLK MFLX06359-72BLK MFLX06359-73BLK MFLX06359-80BLK MFLX06359-70BLK MFLX06359-82BLK MFLX06361-00BLK MFLX06359-81BLK MFLX06359-83BLK
CPC® Quick-Disconnect Fittings, Threaded Bodies
Kiinnitykset ja liittimet
No more struggling - just press thumb latch.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-More Product Information