Union Fittings, Hose Barb, Y-Connector, Polyproylene, Dynalon®
Toimittaja: Avantor Fluid Handling
MFLX06295-10BLKEA 74.2 EUR
MFLX06295-10BLK MFLX06295-20BLK MFLX06295-30BLK MFLX06295-40BLK MFLX06295-50BLK MFLX06295-60BLK MFLX06295-70BLK MFLX06296-00BLK MFLX06296-20BLK MFLX06296-50BLK
Union Fittings, Hose Barb, Y-Connector, Polyproylene, Dynalon®
Kiinnitykset ja liittimet
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for