GF® Signet 316 SS Socket-Weld Mini-Tap Installation Fittings
Toimittaja: Avantor Fluid Handling
05618-62 05618-63
MFLX05618-62EA 828 EUR
MFLX05618-62 MFLX05618-63
GF® Signet 316 SS Socket-Weld Mini-Tap Installation Fittings
Virtausmittarit
Installation fittings for your GF® Signet Paddle Wheel Flowmeters.
  • 316 SS Socket-Weld Mini-Tap Fittings for Metalex™ sensors only
  • Includes cap kit
Fittings are precision crafted to ensure proper sensor insertion depth and accurate flow measurement. Pipe fitting sizes range from 1/2" to 1" in diameter.
Required system components are the flow sensor, installation fitting, and monitor or controller, all sold separately.