Masterflex® Correlated Variable-Area Flowmeters, High-Flow, 150-mm Scale, Avantor®
Toimittaja: Avantor Fluid Handling
03267-87 03267-90 03269-37 03269-86
MFLX03267-32EA 221 EUR
MFLX03267-32 MFLX03267-34 MFLX03267-36 MFLX03269-35 MFLX03269-84 MFLX03267-87 MFLX03267-90 MFLX03269-37 MFLX03269-86
Masterflex® Correlated Variable-Area Flowmeters, High-Flow, 150-mm Scale, Avantor®
Virtausmittarit
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. All correlated flowmeters come with data sheets forMore Product Information