Masterflex® Direct Lift 2- and 3-way Solenoid Valves, Avantor®
Toimittaja: Avantor Fluid Handling
01540-01 01540-02 01540-04 01540-08 01540-09 01540-11 01540-12 01540-13 01540-14 01540-17 01540-18 01540-10
MFLX01540-01EA 160 EUR
MFLX01540-01 MFLX01540-02 MFLX01540-04 MFLX01540-08 MFLX01540-09 MFLX01540-11 MFLX01540-12 MFLX01540-13 MFLX01540-14 MFLX01540-17 MFLX01540-18 MFLX01540-10
Masterflex® Direct Lift 2- and 3-way Solenoid Valves, Avantor®
Venttiilit Solenoidi venttiilit
Ideal for use with high-purity or corrosive fluids.
  • All wetted parts made of inert PTFE
  • Minimal dead volume reduces problems with trapped liquids
Rated for 26" Hg vacuum. Maximum fluid temperature is 158 °F (70 °C). All valves have 15" lead wires. Valves are compact and lightweight — mount them almost anywhere. Valves with a 1/16" orifice have mounting holes at the bottom only and valves with a 3/32" and 5/32" orifice have mounting holes at the top and bottom.