Combined density and refractometry standards
Toimittaja: Mettler - Toledo
51338013 51338014
METT51338013EA 357 EUR
METT51338013 METT51338014
Combined density and refractometry standards
Standardiaineet ja vertailumateriaalit Taitekerroinstandardit
These combined reference standards are intended to reduce uncertainty, eliminate time-consuming and error prone sample transfer and facilitate fully automatic performance verification.