High-density shelving system, Super Erecta™ Top-Track™
Toimittaja: Metro International
TTE21S TTE18S TTM18S TTM24S TTE18K4 TTM18K4
METOTTE21SEA 1764 EUR
METOTTE21S METOTTE18S METOTTM18S METOTTM24S METOTTE18K4 METOTTM18K4
High-density shelving system, Super Erecta™ Top-Track™
Kalusteet Hyllyt
Top-Track™ shelving system increases storage capacity up to 50% and maximises the usable storage area in a given space.
  • Maximum load rating for a single unit is 408,2 kg
  • Stainless steel units provide maximum protection in corrosive environments
The overhead track system guides the mobile units and opens an access aisle between any two units. The track is above the storage units enabling easy access.
Stationary end unit kits include four 2184 mm high posts and the necessary stainless-steel hardware for connecting track sets to two stationary endMore Product Information