Divider tote box for shelving systems, Metro®
Toimittaja: Metro International
TB93050BAS TB93030NAT TB92060BAS TB92035BAS TB93060CAS TB93060NAT
METOTB93050BASEA 119.8 EUR
METOTB93050BAS METOTB93030NAT METOTB92060BAS METOTB92035BAS METOTB93060CAS METOTB93060NAT
Divider tote box for shelving systems, Metro®
Haihdutusmaljat/Tarjottimet/Altaat Alustat, säilytyslokerikot
Divider tote boxes made of polypropylene base resins offer safe and efficient storage of items in bulk, or in compartmentalised fashion with the use of optional dividers and covers. Available in natural gray (NAT), Benstat™ blue static dissipate (BAS) or Bentron™ black conductive (CAS).