Shelving systems, direct wall mounts
Toimittaja: Metro International
1WD18S 2WD18S 1WD14S 1WD14C
METO1WD18SEA 141.5 EUR
METO1WD18S METO2WD18S METO1WD14S METO1WD14C
Shelving systems, direct wall mounts
Kalusteet Hyllyt
Direct wall mount brackets allow the installation of wire or solid shelves directly onto a wall surface. Each brackets consists of one shelf support and mounting plate, with two shelf collar caps. Use single support at shelf ends and double support for adjoining shelves.