Universal ovens, pass-through, with forced air circulation, UF TS series
Toimittaja: MEMMERT
UF260TS UF450TS UF750TS UF160TS
MEMMUF260TSEA 11250 EUR
MEMMUF260TS MEMMUF450TS MEMMUF750TS MEMMUF160TS
Universal ovens, pass-through, with forced air circulation, UF TS series
Lämpökaapit Uunit, puhallinavusteinen ja luonnollinen ilmankierto
The UF TS pass through ovens help to minimise contamination when transporting samples or components. They have an easy-to-clean interior, fast recovery times after opening the door as well as homogeneous temperature distribution, this oven is the perfect material lock between the grey room and the cleanroom in the electronics, semiconductor, medicine or pharmaceutical industries.
  • Intuitive and easy-to-use operating menu
  • Air exchange rates and air flap position are electronically controllable
  • Temperature and air exchange ramps are programmable via