Bottom drip trays for IN/INplus and IF/IFplus incubators
Toimittaja: MEMMERT
B29722IF260 B29722IF260PLS B29722IN260 B29722IN260PLS B04356IF30 B04356IF30PLUS B04356IN30 B04356IN30PLUS B04359IF160 B04358IF55 B04358IF75 B04358IF75PLUS B04358IF55PLUS B04358IN55 B04358IN55PLUS B04358IN75 B04358IN75PLUS B04359IF110 B04359IF110PLS B04359IF160PLS B04359IN110 B04359IN110PLS B04359IN160 B04359IN160PLS B04362IF450 B04362IF450PLS B04362IF750 B04362IF750PLS B04362IN450 B04362IN450PLS B04362IN750 B04362IN750PLS
MEMMB29722IF260EA 256 EUR
MEMMB29722IF260 MEMMB29722IF260PLS MEMMB29722IN260 MEMMB29722IN260PLS MEMMB04356IF30 MEMMB04356IF30PLUS MEMMB04356IN30 MEMMB04356IN30PLUS MEMMB04359IF160 MEMMB04358IF55 MEMMB04358IF75 MEMMB04358IF75PLUS MEMMB04358IF55PLUS MEMMB04358IN55 MEMMB04358IN55PLUS MEMMB04358IN75 MEMMB04358IN75PLUS MEMMB04359IF110 MEMMB04359IF110PLS MEMMB04359IF160PLS MEMMB04359IN110 MEMMB04359IN110PLS MEMMB04359IN160 MEMMB04359IN160PLS MEMMB04362IF450 MEMMB04362IF450PLS MEMMB04362IF750 MEMMB04362IF750PLS MEMMB04362IN450 MEMMB04362IN450PLS MEMMB04362IN750 MEMMB04362IN750PLS
Bottom drip trays for IN/INplus and IF/IFplus incubators
Haihdutusmaljat/Tarjottimet/Altaat Roiskealustat/-astiat
May affect the temperature distribution in the chamber.